accessibility menu, dialog, popup

UserName

CAS RN 4522-93-4

H3COOFFFFF

CAS RN 4522-93-4

IUPAC Name: ethyl 2,3,4,5,6-pentafluorobenzoate
Synonyms: ethyl 2,3,4,5,6-pentafluorobenzoate
Molecular Weight (g/mol): 240.13
Molecular Formula: C9H5F5O2
InChi Key: DFUDMSIRGGTHGI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results