accessibility menu, dialog, popup

UserName

CAS RN 455-38-9

HOOF

CAS RN 455-38-9

IUPAC Name: 3-fluorobenzoic acid
Synonyms: 3-fluorobenzoic acid M-fluorobenzoic acid
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: MXNBDFWNYRNIBH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(F)=C1
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results