accessibility menu, dialog, popup

UserName

CH3FNOO

CAS RN 455-88-9

IUPAC Name: 1-fluoro-2-methyl-4-nitrobenzene
Synonyms: 1-fluoro-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: XUCYJGMIICONES-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1F)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results