accessibility menu, dialog, popup

UserName

CAS RN 45791-36-4

IUPAC Name: 1-(4-bromophenyl)ethan-1-amine
Synonyms: 1-(4-bromophenyl)ethanamine
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: SOZMSEPDYJGBEK-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results