accessibility menu, dialog, popup

UserName

NH2SNHF

CAS RN 458-05-9

IUPAC Name: (3-fluorophenyl)thiourea
Synonyms: 3-fluorophenylthiourea
Molecular Weight (g/mol): 170.21
Molecular Formula: C7H7FN2S
InChi Key: YKOBLHQALWQKTC-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC(F)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results