accessibility menu, dialog, popup

UserName

CAS RN 459-31-4

IUPAC Name: 3-(4-fluorophenyl)propanoate
Synonyms: 3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 167.16
Molecular Formula: C9H8FO2
InChi Key: ZMKXWDPUXLPHCA-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results