CAS RN 460085-45-4
CAS RN 460085-45-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methylumbelliferyl 2-Sulfamino-2-deoxy-Alpha-D-glucopyranoside Sodium Salt, TRC
CAS: 460085-45-4 Molecular Formula: C16 H18 N O10 S . Na Molecular Weight (g/mol): 439.37 Synonym: 7-[[2-Deoxy-2-(sulfoamino)-α-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one Sodium Salt; IUPAC Name: sodium;N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]sulfamate SMILES: [Na+].CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3NS(=O)(=O)[O-])ccc12