missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2-Sulfamino-2-deoxy-Alpha-D-glucopyranoside Sodium Salt, TRC

Product Code. 30007990
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007990 5 mg 5mg
30003621 10 mg 10mg
30007539 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007990 Supplier Toronto Research Chemicals Supplier No. TRCM3348515MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460085-45-4
Molecular Formula C16 H18 N O10 S . Na
Molecular Weight (g/mol) 439.37
Synonym 7-[[2-Deoxy-2-(sulfoamino)-α-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one Sodium Salt;
IUPAC Name sodium;N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]sulfamate
SMILES [Na+].CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3NS(=O)(=O)[O-])ccc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H19NO10S.Na/c1-7-4-12(19)26-10-5-8(2-3-9(7)10)25-16-13(17-28(22,23)24)15(21)14(20)11(6-18)27-16;/h2-5,11,13-18,20-21H,6H2,1H3,(H,22,23,24);/q;+1/p-1/t11-,13-,14-,15-,16+;/m1./s1
Formula Weight 439.0549
Percent Purity >95%
Chemical Name or Material 4-Methylumbelliferyl 2-Sulfamino-2-deoxy-alpha-D-glucopyranoside Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.