accessibility menu, dialog, popup

UserName

CAS RN 461-89-2

IUPAC Name: 2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
Synonyms: azauracil s-triazine-3,5(2H,4H)-dione 4(6)-azauracil 1,2,4-triazine-3,5-diol as-triazine-3,5-diol
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: SSPYSWLZOPCOLO-UHFFFAOYSA-N
SMILES: O=C1NN=CC(=O)N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)