missing translation for 'onlineSavingsMsg'
Learn More

6-Azauracil, 98%

Product Code. 11496987
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496987 5 g 5g
11406997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496987 Supplier Thermo Scientific Alfa Aesar Supplier No. A14389.06
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 461-89-2 | C3H3N3O2 | 113.08 g/mol

6-Azauracil (6-AU) is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Azauracil (6-AU) is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.

Solubility
Partly soluble in water, ethanol, DMSO, and 1 M NH4OH (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-89-2
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00006456
InChI Key SSPYSWLZOPCOLO-UHFFFAOYSA-N
Synonym 6-azauracil, 1,2,4-triazine-3,5 2h,4h-dione, azauracil, 1,2,4-triazine-3,5-diol, 4 6-azauracil, as-triazine-3,5-diol, as-triazine-3,5 2h,4h-dione, 1,2,4-triazine-3,5-dione, usaf cb-30, unii-i14twn70lr
PubChem CID 68037
ChEBI CHEBI:53745
IUPAC Name 2H-1,2,4-triazine-3,5-dione
SMILES O=C1NN=CC(=O)N1

Specifications

Melting Point 274°C to 282°C
Quantity 5 g
Beilstein 116472
Solubility Information Partly soluble in water,ethanol,DMSO,and 1 M NH4OH (50mg/ml).
Formula Weight 113.08
Percent Purity 98%
Chemical Name or Material 6-Azauracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.