accessibility menu, dialog, popup

UserName

FFBr

CAS RN 461-96-1

IUPAC Name: 1-bromo-3,5-difluorobenzene
Synonyms: 1-bromo-3,5-difluorobenzene
Molecular Weight (g/mol): 192.99
Molecular Formula: C6H3BrF2
InChi Key: JHLKSIOJYMGSMB-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
  • (2)
Boiling Point 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)