accessibility menu, dialog, popup

UserName

CH3BrBrHOBr

CAS RN 4619-74-3

IUPAC Name: 2,4,6-tribromo-3-methylphenol
Synonyms: tribromometacresol
Molecular Weight (g/mol): 344.83
Molecular Formula: C7H5Br3O
InChi Key: QKHROXOPRBWBDD-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(Br)C(O)=C1Br
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results