accessibility menu, dialog, popup

UserName

CH3OCH3O(Z)

CAS RN 4652-27-1

IUPAC Name: (3Z)-4-methoxybut-3-en-2-one
Synonyms: (3Z)-4-methoxybut-3-en-2-one
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: VLLHEPHWWIDUSS-ARJAWSKDSA-N
SMILES: CO\C=C/C(C)=O
Boiling Point 
  • (2)
  • (3)
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results