missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-buten-2-one, 90%, tech.

Product Code. 10103590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10103590 50 mL Glass bottle 50mL
10103540 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103590 Supplier Thermo Scientific Acros Supplier No. 153390500
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4652-27-1 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4652-27-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008778,MFCD00008778,MFCD00008778
InChI Key VLLHEPHWWIDUSS-ARJAWSKDSA-N
Synonym z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
PubChem CID 643837
IUPAC Name (Z)-4-methoxybut-3-en-2-one
SMILES CO\C=C/C(C)=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 200°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH=CHCOCH3
Refractive Index 1.4650 to 1.469
Quantity 50 mL
Beilstein 01, IV, 4080
Fieser 10,258
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Formula Weight 100.12
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Methoxy-3-buten-2-one, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-3-buten-2-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.