CAS RN 470-69-9
CAS RN 470-69-9
IUPAC Name:
(2R,3R,4S,5S,6R)-2-{[(2S,3S,5R)-2-({[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}methyl)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
(2R,3R,4S,5S,6R)-2-{[(2S,3S,5R)-2-({[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}methyl)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
504.44
Molecular Formula:
C18H32O16
InChi Key:
VAWYEUIPHLMNNF-CMCDSJAFSA-N
SMILES:
OC[C@H]1O[C@@](CO)(OC[C@@]2(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)O[C@H](CO)C(O)[C@@H]2O)[C@@H](O)[C@@H]1O
1
–
1
of
1
results
1
1-Kestose, MedChemExpress
MedChemExpress 1-Kestose, the smallest fructooligosaccharide component, which efficiently stimulates Faecalibacterium prausnitzii as well as Bifidobacteria.