CAS RN 4707-50-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Divarinic Acid, TRC
CAS: 4707-50-0 Molecular Formula: C10H12O4 Molecular Weight (g/mol): 196.2 Synonym: 6-Propyl-b-resorcylic Acid, (7CI,8CI),2,4-Dihydroxy-6-propylbenzoic Acid,Divaric Acid SMILES: OC(C(C(O)=CC(O)=C1)=C1CCC)=O