CAS RN 475060-00-5
CAS RN 475060-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methoxy-4-methyl-2-pyridinamine, TRC
CAS: 475060-00-5 Molecular Formula: C7H10N2O Molecular Weight (g/mol): 138.17 Synonym: 3-Methoxy-4-methylpyridin-2-amine IUPAC Name: 3-methoxy-4-methylpyridin-2-amine SMILES: COc1c(C)ccnc1N