missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-methyl-2-pyridinamine, TRC

Product Code. 30476275
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476275 50 mg 50mg
30467903 500 mg 500mg
30472238 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476275 Supplier Toronto Research Chemicals Supplier No. TRCM33062350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475060-00-5
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
Synonym 3-Methoxy-4-methylpyridin-2-amine
IUPAC Name 3-methoxy-4-methylpyridin-2-amine
SMILES COc1c(C)ccnc1N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H10N2O/c1-5-3-4-9-7(8)6(5)10-2/h3-4H,1-2H3,(H2,8,9)
Formula Weight 138.0793
Chemical Name or Material 3-Methoxy-4-methyl-2-pyridinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.