accessibility menu, dialog, popup

UserName

CH3OOOOOOOH3COOH3CBrOOCH3OOCH3OOH3COOCH3

CAS RN 4753-07-5

IUPAC Name: [3,4,5-tris(acetyloxy)-6-{[4,5-bis(acetyloxy)-2-[(acetyloxy)methyl]-6-bromooxan-3-yl]oxy}oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-{[4,5-bis(acetyloxy)-2-[(acetyloxy)methyl]-6-bromooxan-3-yl]oxy}oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 699.45
Molecular Formula: C26H35BrO17
InChi Key: NLFHLQWXGDPOME-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(Br)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)