accessibility menu, dialog, popup

UserName

H3CSnH3CCH3N

CAS RN 478282-21-2

IUPAC Name: 8-(tributylstannyl)quinoline
Synonyms: 8-(tributylstannyl)quinoline
Molecular Weight (g/mol): 418.21
Molecular Formula: C21H33NSn
InChi Key: CFXBLYUOIPJSEG-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=C2N=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results