missing translation for 'onlineSavingsMsg'
Learn More

8-(Tri-n-butylstannyl)quinoline, 95%, Thermo Scientific™

Product Code. 15488097
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488097 250 mg 250mg
15478097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488097 Supplier Thermo Scientific Alfa Aesar Supplier No. H51471.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 478282-21-2
Molecular Formula C21H33NSn
Molecular Weight (g/mol) 418.21
MDL Number MFCD09265482
InChI Key CFXBLYUOIPJSEG-UHFFFAOYSA-N
Synonym 8-tributylstannyl quinoline, quinoline-8-tributylstannane, quinoline, 8-tributylstannyl, 8-tri-n-butylstannyl quinoline
PubChem CID 12179403
SMILES CCCC[Sn](CCCC)(CCCC)C1=C2N=CC=CC2=CC=C1

Specifications

Quantity 250 mg
UN Number UN2788
IUPAC Name tributyl(quinolin-8-yl)stannane
Formula Weight 418.19
Percent Purity 95%
Chemical Name or Material 8-(Tri-n-butylstannyl)quinoline
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 8-(Tri-n-butylstannyl)quinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 8-(Tri-n-butylstannyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.