accessibility menu, dialog, popup

UserName

ClSOOCl

CAS RN 478647-00-6

IUPAC Name: 3'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 3'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 287.15
Molecular Formula: C12H8Cl2O2S
InChi Key: FVELIXWAKJVRDG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results