CAS RN 479-50-5
CAS RN 479-50-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lucanthone, MedChemExpress
MedChemExpress Lucanthone is an endonuclease inhibitor of Apurinic endonuclease-1 (APE-1).