missing translation for 'onlineSavingsMsg'
Learn More

Lucanthone, MedChemExpress

Product Code. 16452369
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452369 1 mg 1mg
16470616 20 mg 20mg
16431148 5 mg 5mg
16418738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452369 Supplier Medchem Express Supplier No. HYB2098
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Lucanthone is an endonuclease inhibitor of Apurinic endonuclease-1 (APE-1).

Lucanthone is an endonuclease inhibitor of Apurinic endonuclease-1 (APE-1).

Specifications

Chemical Name or Material Lucanthone
CAS 479-50-5
Color Orange
Purity Grade Notes Research
Molecular Formula C20H24N2OS
Quantity 1 mg
Solubility Information DMSO : 25 mg/mL (73.43 mM; Need ultrasonic)
SMILES O=C1C2=C(SC3=C1C=CC=C3)C(C)=CC=C2NCCN(CC)CC
Molecular Weight (g/mol) 340.48
Formula Weight 340.48
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.