accessibility menu, dialog, popup

UserName

CAS RN 480-41-1

HOOOHOHO

CAS RN 480-41-1

IUPAC Name: 5,7-dihydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: asahina naringenine pelargidanon naringetol salipurpol salipurol narigenin
Molecular Weight (g/mol): 272.26
Molecular Formula: C15H12O5
InChi Key: FTVWIRXFELQLPI-UHFFFAOYNA-N
SMILES: OC1=CC=C(C=C1)C1CC(=O)C2=C(O)C=C(O)C=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results