accessibility menu, dialog, popup

UserName

CH3OH3CBrOCH3

CAS RN 480439-43-8

IUPAC Name: 1-bromo-3-(1,1-diethoxyethyl)benzene
Synonyms: 1-bromo-3-(1,1-diethoxyethyl)benzene
Molecular Weight (g/mol): 273.17
Molecular Formula: C12H17BrO2
InChi Key: XKQAVDMOLWVSRW-UHFFFAOYSA-N
SMILES: CCOC(C)(OCC)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results