accessibility menu, dialog, popup

UserName

CH3FFFSOOOFFFSOOOCuCu

CAS RN 48209-28-5

IUPAC Name: bis(λ¹-copper(1+)) toluene ditrifluoromethanesulfonate
Synonyms: bis(λ¹-copper(1+)) toluene ditriflate
Molecular Weight (g/mol): 517.36
Molecular Formula: C9H8Cu2F6O6S2
InChi Key: ATUUSMBCJWYMEG-UHFFFAOYSA-L
SMILES: [Cu+].[Cu+].CC1=CC=CC=C1.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results