Learn More
Copper(I) trifluoromethanesulfonate toluene complex (2:1), 98%
Description
Copper(I) trifluoromethanesulfonate toluene complex serves as a vital coupling reagent for photo reactions. It is also used in the preparation of polycyclic furans and butyrolactones. Further, it is used as a reagent for the preparation of beta-lactam from beta-amino thiol-esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(I) trifluoromethanesulfonate toluene complex serves as a vital coupling reagent for photo reactions. It is also used in the preparation of polycyclic furans and butyrolactones. Further, it is used as a reagent for the preparation of beta-lactam from beta-amino thiol-esters.
Solubility
Soluble in toluene.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 180°C (decomposition) |
| Flash Point | 76°C (168°F) |
| Quantity | 5 g |
| UN Number | UN1325 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in toluene. |
| Formula Weight | 517.37 |
| Percent Purity | 98% |
| Chemical Name or Material | Copper(I) trifluoromethanesulfonate toluene complex (2:1) |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.