accessibility menu, dialog, popup

UserName

CAS RN 489-01-0

H3COOHH3CH3CCH3CH3H3CCH3

CAS RN 489-01-0

IUPAC Name: 2,6-di-tert-butyl-4-methoxyphenol
Synonyms: 2,6-di-tert-butyl-4-methoxyphenol
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: SLUKQUGVTITNSY-UHFFFAOYSA-N
SMILES: COC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

 No Refinements