accessibility menu, dialog, popup

UserName

CAS RN 490-64-2

H3COOCH3OCH3OO

CAS RN 490-64-2

IUPAC Name: 2,4,5-trimethoxybenzoate
Synonyms: 2,4,5-trimethoxybenzoate
Molecular Weight (g/mol): 211.19
Molecular Formula: C10H11O5
InChi Key: KVZUCOGWKYOPID-UHFFFAOYSA-M
SMILES: COC1=CC(OC)=C(C=C1OC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results