accessibility menu, dialog, popup

UserName

CAS RN 491-30-5

IUPAC Name: 1,2-dihydroisoquinolin-1-one
Synonyms: isocarbostyril isoquinolin-1-one isocarbostyril (van) (8CI)
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: VDBNYAPERZTOOF-UHFFFAOYSA-N
SMILES: O=C1NC=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)