missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyisoquinoline, 97%

Product Code. 11442240
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442240 10 g 10g
11432240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442240 Supplier Thermo Scientific Alfa Aesar Supplier No. H29124.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 491-30-5 | C9H7NO | 145.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-30-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00006899
InChI Key VDBNYAPERZTOOF-UHFFFAOYSA-N
Synonym 1-hydroxyisoquinoline, isoquinolin-1 2h-one, isocarbostyril, 1-isoquinolinol, 1 2h-isoquinolinone, isoquinolin-1-ol, isoquinolin-1-one, 1 2h-isoquinolone, 1-hydroxyisoquinolin, isoquinolinol
PubChem CID 10284
ChEBI CHEBI:18350
IUPAC Name 2H-isoquinolin-1-one
SMILES C1=CC=C2C(=C1)C=CNC2=O

Specifications

Melting Point 212°C to 215°C
Quantity 10 g
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material 1-Hydroxyisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.