accessibility menu, dialog, popup

UserName

OOOO

CAS RN 492-94-4

IUPAC Name: bis(furan-2-yl)ethane-1,2-dione
Synonyms: α-furil furan, 2-coco-furyl
Molecular Weight (g/mol): 190.15
Molecular Formula: C10H6O4
InChi Key: SXPUVBFQXJHYNS-UHFFFAOYSA-N
SMILES: O=C(C(=O)C1=CC=CO1)C1=CC=CO1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)