missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furil, 98%

Product Code. 11488336
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488336 100 g 100g
11478336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488336 Supplier Thermo Scientific Alfa Aesar Supplier No. A15386.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 492-94-4 | C10H6O4 | 190.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-94-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00003241
InChI Key SXPUVBFQXJHYNS-UHFFFAOYSA-N
Synonym furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
PubChem CID 68119
IUPAC Name 1,2-bis(furan-2-yl)ethane-1,2-dione
SMILES C1=COC(=C1)C(=O)C(=O)C2=CC=CO2

Specifications

Melting Point 162°C to 165°C
Quantity 100 g
Beilstein 383882
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material 2,2'-Furil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.