accessibility menu, dialog, popup

UserName

OOOO

CAS RN 4940-39-0

IUPAC Name: 4-oxo-4H-chromene-2-carboxylate
Synonyms: 4-oxochromene-2-carboxylate
Molecular Weight (g/mol): 189.15
Molecular Formula: C10H5O4
InChi Key: RVMGXWBCQGAWBR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=O)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results