missing translation for 'onlineSavingsMsg'
Learn More

Chromone-2-carboxylic acid, 97%

Product Code. 11440044
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440044 25 g 25g
11430044 5 g 5g
11450044 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440044 Supplier Thermo Scientific Alfa Aesar Supplier No. A13552.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4940-39-0 | C10H6O4 | 190.154 g/mol

Chromone-2-carboxylic acid is used in the cyclic form 3?-oxopyrazolidino[4?,5??2, 3]-chroman-4-one, gives the azide of chromone-2-carboxylic acid, the curtius rearrangement of which is used to synthesize a number of 2-acylaminochromones and it is an orally active antiallergic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromone-2-carboxylic acid is used in the cyclic form 3′-oxopyrazolidino[4′,5′∶2, 3]-chroman-4-one, gives the azide of chromone-2-carboxylic acid, the curtius rearrangement of which is used to synthesize a number of 2-acylaminochromones and it is an orally active antiallergic agent.

Solubility
Soluble in alcohol, water, DMSO, hot methanol, and ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4940-39-0
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006838
InChI Key RVMGXWBCQGAWBR-UHFFFAOYSA-N
Synonym chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
PubChem CID 2741
IUPAC Name 4-oxochromene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O

Specifications

Melting Point ∼260°C (decomposition)
Quantity 25 g
Beilstein 146442
Merck Index 14,2237
Solubility Information Soluble in alcohol,water,DMSO,hot methanol,and ethanol.
Formula Weight 190.15
Percent Purity 97%
Chemical Name or Material Chromone-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.