accessibility menu, dialog, popup

UserName

NH2SNHCl

CAS RN 4947-89-1

IUPAC Name: (3-chlorophenyl)thiourea
Synonyms: 3-chlorophenylthiourea
Molecular Weight (g/mol): 186.66
Molecular Formula: C7H7ClN2S
InChi Key: LSYZRUOXXOTVAV-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results