accessibility menu, dialog, popup

UserName

HOOO

CAS RN 496-41-3

IUPAC Name: 1-benzofuran-2-carboxylic acid
Synonyms: 2-benzofurancarboxylic acid
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: OFFSPAZVIVZPHU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CC=C2O1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results