missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxylic acid, 98+%

Product Code. 11483057
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483057 25 g 25g
11463057 1 g 1g
11473057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483057 Supplier Thermo Scientific Alfa Aesar Supplier No. A15579.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 496-41-3 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-41-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00005848
InChI Key OFFSPAZVIVZPHU-UHFFFAOYSA-N
Synonym benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
PubChem CID 10331
IUPAC Name 1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2O1

Specifications

Melting Point 192°C to 196°C
Boiling Point 310°C to 315°C
Quantity 25 g
Beilstein 124204
Merck Index 14,2561
Formula Weight 162.15
Percent Purity ≥98%
Chemical Name or Material Benzo[b]furan-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.