accessibility menu, dialog, popup

UserName

CAS RN 496-69-5

OHFBr

CAS RN 496-69-5

IUPAC Name: 2-bromo-4-fluorophenol
Synonyms: 2-bromo-4-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: MEYRABVEYCFHHB-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1Br
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)