accessibility menu, dialog, popup

UserName

BrOO

CAS RN 49619-82-1

IUPAC Name: 3-bromo-4H-chromen-4-one
Synonyms: 3-bromochromen-4-one
Molecular Weight (g/mol): 225.04
Molecular Formula: C9H5BrO2
InChi Key: IQIGYNPOESZBDJ-UHFFFAOYSA-N
SMILES: BrC1=COC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results