missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4H-chromen-4-one, 97%, Thermo Scientific™

Product Code. 10631873
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10631873 1 g 1g
10260215 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10631873 Supplier Thermo Scientific Maybridge Supplier No. RF01262DA
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 49619-82-1
Molecular Formula C9H5BrO2
Molecular Weight (g/mol) 225.041
MDL Number MFCD00017337
InChI Key IQIGYNPOESZBDJ-UHFFFAOYSA-N
Synonym 3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
PubChem CID 521256
IUPAC Name 3-bromochromen-4-one
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 225.04
Percent Purity 97%
Chemical Name or Material 3-Bromo-4H-chromen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.