accessibility menu, dialog, popup

UserName

OHONOHO

CAS RN 499-83-2

IUPAC Name: pyridine-2,6-dicarboxylic acid
Synonyms: dipicolinic acid dipicolinic acid, monosodium salt dipicolinic acid, disodium salt dipotassium salt dipicolinic acid zinc dipicolinate dipicolinic acid, dipotassium salt disodium salt dipicolinic acid monosodium salt dipicolinic acid zinc salt dipicolinic acid 2,6-dipicolinic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: WJJMNDUMQPNECX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=N1)C(O)=O
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)