missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxylic Acid, TraceSELECT™, ≥99.999% (metals basis), Solstice

Product Code. 15617520 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15617520 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15617520 Supplier Honeywell-Fluka Supplier No. 9425625G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 499-83-2
Color White
Packaging Plastic bottle
Molecular Formula C7H5NO4
MDL Number MFCD00006299
Quantity 25 g
UN Number NONH for all modes of transport
Beilstein 131629
Synonym 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
InChI Key WJJMNDUMQPNECX-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1)C(=O)O)C(=O)O
IUPAC Name pyridine-2,6-dicarboxylic acid
Molecular Weight (g/mol) 167.12
PubChem CID 10367
ChEBI CHEBI:46837
Formula Weight 167.12g/mol
Percent Purity ≥99.999% (metals basis)
Grade TraceSELECT
Physical Form Crystalline
Chemical Name or Material 2,6-Pyridinedicarboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.