CAS RN 499770-76-2
CAS RN 499770-76-2
IUPAC Name:
5-(bromomethyl)-1-methyl-1H-1,2,3-benzotriazole
Synonyms:
5-(bromomethyl)-1-methyl-1,2,3-benzotriazole
Molecular Weight (g/mol):
226.08
Molecular Formula:
C8H8BrN3
InChi Key:
OSUZHHPMRAIJDY-UHFFFAOYSA-N
SMILES:
CN1N=NC2=CC(CBr)=CC=C12
1
–
1
of
1
results
1
5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™
CAS: 499770-76-2 Molecular Formula: C8H8BrN3 Molecular Weight (g/mol): 226.077 InChI Key: OSUZHHPMRAIJDY-UHFFFAOYSA-N Synonym: 5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole,5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole,5-bromomethyl-1-methylbenzotriazole,5-bromomethyl-1-methyl-benzotriazole,1h-benzotriazole, 5-bromomethyl-1-methyl,5-bromomethyl-1-methyl-1,2,3-benzotriazole,5-bromomethyl-1-methyl-1h-benzotriazole PubChem CID: 2795418 IUPAC Name: 5-(bromomethyl)-1-methylbenzotriazole SMILES: CN1C2=C(C=C(C=C2)CBr)N=N1