missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™

Product Code. 10672495
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10672495 5 g Amber Glass Bottle 5g
10600913 250 mg Amber Glass Bottle 250mg
10435892 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10672495 Supplier Thermo Scientific Maybridge Supplier No. CC23508DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-76-2
Molecular Formula C8H8BrN3
Molecular Weight (g/mol) 226.077
InChI Key OSUZHHPMRAIJDY-UHFFFAOYSA-N
Synonym 5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole
PubChem CID 2795418
IUPAC Name 5-(bromomethyl)-1-methylbenzotriazole
SMILES CN1C2=C(C=C(C=C2)CBr)N=N1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 226.08
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.