accessibility menu, dialog, popup

UserName

CH3NNNFFFOOH

CAS RN 499771-21-0

IUPAC Name: 5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid
Synonyms: 5-methyl-1-[3-(trifluoromethyl)phenyl]-1,2,3-triazole-4-carboxylic acid
Molecular Weight (g/mol): 271.2
Molecular Formula: C11H8F3N3O2
InChi Key: IRJLZHQBWAUKGA-UHFFFAOYSA-N
SMILES: CC1=C(N=NN1C1=CC=CC(=C1)C(F)(F)F)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results