accessibility menu, dialog, popup

UserName

CAS RN 501945-71-7

IUPAC Name: 2-(1-benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(1-benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 260.16
Molecular Formula: C14H17BO2S
InChi Key: YFTHTJAPODJVSL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results