missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Thermo Scientific™

Product Code. 10776905
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10776905 5 g Amber Glass Bottle 5g
10341613 250 mg Amber Glass Bottle 250mg
10187352 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10776905 Supplier Thermo Scientific Maybridge Supplier No. CC24839DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 501945-71-7
Molecular Formula C14H17BO2S
Molecular Weight (g/mol) 260.16
MDL Number MFCD05664405
InChI Key YFTHTJAPODJVSL-UHFFFAOYSA-N
Synonym 2-benzo b thiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzothiophene-5-boronic acid pinacol ester, 1-benzothiophen-5-yl-4,4,5,5tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-benzo b thien-5-yl-4,4,5,5-tetramethyl, 1-benzothiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-benzo b thiophen-5-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo b thiophene, 2-1-benzothiophen-5-yl-4,4,5,5-tetra-methyl-1,3,2-dioxaborolane, benzo b thiophene,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl
PubChem CID 2795455
IUPAC Name 2-(1-benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C2SC=CC2=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 260.16
Percent Purity 95%
Chemical Name or Material 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.