accessibility menu, dialog, popup

UserName

H3COOSOOCH3

CAS RN 50397-64-3

IUPAC Name: methyl 2-(4-methylbenzenesulfonyl)acetate
Synonyms: methyl 2-(4-methylbenzenesulfonyl)acetate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: JMUZMDKWJWSBCU-UHFFFAOYSA-N
SMILES: COC(=O)CS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results